Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrpl45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105834.1 | NP_001099304.1 | 7 - 8 | 876 | 87 | |
GenBank mRNA | FQ216878.1 | - | 6 - 7 | 668 | 87 | |
FQ225351.1 | - | 6 - 7 | 670 | 87 | ||
FQ226279.1 | - | 6 - 7 | 668 | 87 | ||
FQ229232.1 | - | 6 - 7 | 666 | 87 | ||
FQ232005.1 | - | 6 - 7 | 669 | 87 | ||
FQ233183.1 | - | 6 - 7 | 668 | 87 | ||
FQ233508.1 | - | 6 - 7 | 668 | 87 | ||
FQ233844.1 | - | 6 - 7 | 668 | 87 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |