Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myocd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_182667.2 | NP_872608.1 | 10 - 11 | 2296 | 120 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protein Kinase Rattus norvegicus Probe spans exons |