Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpl23a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108283.1 | NP_001101753.1 | 4 - 5 | 497 | 95 | |
GenBank mRNA | BC168782.1 | - | 4 - 5 | 472 | 6921912 | 95 |
FQ210401.1 | - | 4 - 5 | 472 | 95 | ||
FQ210899.1 | - | 4 - 5 | 473 | 95 | ||
FQ210939.1 | - | 4 - 5 | 473 | 95 | ||
FQ212155.1 | - | 4 - 5 | 472 | 95 | ||
FQ213974.1 | - | 4 - 5 | 472 | 95 | ||
FQ220609.1 | - | 4 - 5 | 472 | 95 | ||
FQ221032.1 | - | 4 - 5 | 472 | 95 | ||
FQ221170.1 | - | 4 - 5 | 463 | 95 | ||
FQ221186.1 | - | 4 - 5 | 472 | 95 | ||
FQ222102.1 | - | 4 - 5 | 472 | 95 | ||
FQ222310.1 | - | 4 - 5 | 472 | 95 | ||
FQ222471.1 | - | 4 - 5 | 472 | 95 | ||
FQ222491.1 | - | 4 - 5 | 472 | 95 | ||
FQ222732.1 | - | 4 - 5 | 473 | 95 | ||
FQ223377.1 | - | 4 - 5 | 472 | 95 | ||
FQ223511.1 | - | 2 - 3 | 188 | 95 | ||
FQ224628.1 | - | 4 - 5 | 472 | 95 | ||
FQ228299.1 | - | 4 - 5 | 476 | 95 | ||
FQ228395.1 | - | 4 - 5 | 470 | 95 | ||
FQ228575.1 | - | 4 - 5 | 472 | 95 | ||
FQ229110.1 | - | 4 - 5 | 472 | 95 | ||
X65228.1 | - | 4 - 5 | 475 | 95 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Probe spans exons |