Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prss33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109027.1 | NP_001102497.1 | 3 - 4 | 253 | 92 | |
XM_006245874.2 | XP_006245936.1 | 4 - 5 | 543 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Protease Detect Genomic DNA Rattus norvegicus Probe spans exons |