Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B3gntl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015035.1 | NP_001015035.1 | 10 - 11 | 1029 | 72 | |
XM_006247904.2 | XP_006247966.1 | 10 - 11 | 1079 | 72 | ||
XM_008768505.1 | XP_008766727.1 | 10 - 11 | 1079 | 72 | ||
XM_008768506.1 | XP_008766728.1 | 10 - 11 | 1079 | 72 | ||
XM_008768507.1 | XP_008766729.1 | 10 - 11 | 1079 | 72 | ||
XM_008768508.1 | XP_008766730.1 | 10 - 11 | 1079 | 72 | ||
GenBank mRNA | AY634365.1 | - | 10 - 11 | 1029 | 72 | |
BC111711.1 | - | 11 - 12 | 1078 | 7383845 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |