Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B3gntl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015035.1 | NP_001015035.1 | 2 - 3 | 271 | 90 | |
XM_006247904.2 | XP_006247966.1 | 2 - 3 | 321 | 90 | ||
XM_008768503.1 | XP_008766725.1 | 2 - 3 | 321 | 90 | ||
XM_008768504.1 | XP_008766726.1 | 2 - 3 | 321 | 90 | ||
XM_008768505.1 | XP_008766727.1 | 2 - 3 | 321 | 90 | ||
XM_008768506.1 | XP_008766728.1 | 2 - 3 | 321 | 90 | ||
XM_008768507.1 | XP_008766729.1 | 2 - 3 | 321 | 90 | ||
XM_008768508.1 | XP_008766730.1 | 2 - 3 | 321 | 90 | ||
XM_008768509.1 | XP_008766731.1 | 2 - 3 | 321 | 90 | ||
XM_008768510.1 | XP_008766732.1 | 1 - 2 | 89 | 90 | ||
GenBank mRNA | AY634365.1 | - | 2 - 3 | 271 | 90 | |
BC111711.1 | - | 2 - 3 | 245 | 7383845 | 90 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |