Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Afmid | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001111366.1 | NP_001104836.1 | 6 - 7 | 487 | 67 | |
XM_006247823.2 | XP_006247885.1 | 6 - 7 | 503 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Probe spans exons |