Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107045.1 | NP_001100515.1 | 3 - 4 | 347 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |