Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107045.1 | NP_001100515.1 | 4 - 5 | 1315 | 89 | |
XM_006247303.2 | XP_006247365.1 | 3 - 4 | 1077 | 89 | ||
GenBank mRNA | BC168669.1 | - | 3 - 4 | 1083 | 7934743 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |