Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pycr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105857.2 | NP_001099327.1 | 3 - 4 | 322 | 61 | |
XM_008768472.1 | XP_008766694.1 | 4 - 5 | 519 | 61 | ||
XM_008768473.1 | XP_008766695.1 | 4 - 5 | 453 | 61 | ||
XM_008768474.1 | XP_008766696.1 | 4 - 5 | 443 | 61 | ||
XM_008768475.1 | XP_008766697.1 | 4 - 5 | 519 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Probe spans exons |