Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myh8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100485.1 | NP_001093955.1 | 1971 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Rattus norvegicus Probe spans exons |