Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxw11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106993.1 | NP_001100463.1 | 2 - 3 | 244 | 69 | |
XM_006246092.2 | XP_006246154.1 | 2 - 3 | 273 | 69 | ||
XM_006246093.2 | XP_006246155.1 | 2 - 3 | 273 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons |