Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmc8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126301.1 | NP_001119773.1 | 3 - 4 | 488 | 69 | |
XM_006247818.2 | XP_006247880.1 | 3 - 4 | 936 | 69 | ||
XM_006247819.2 | XP_006247881.1 | 3 - 4 | 936 | 69 | ||
XM_006247821.2 | XP_006247883.1 | 3 - 4 | 470 | 69 | ||
XM_008768392.1 | XP_008766614.1 | 3 - 4 | 461 | 69 | ||
GenBank mRNA | BC160873.1 | - | 3 - 4 | 488 | 7366375 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |