Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ube2z | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037643.2 | NP_001032732.2 | 4 - 5 | 741 | 93 | |
GenBank mRNA | BC107663.1 | - | 4 - 5 | 741 | 7936709 | 93 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Apoptosis Mammalian Gene Collection Rattus norvegicus Probe spans exons |