Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hbz | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001172845.1 | NP_001166316.1 | 2 - 3 | 349 | 73 | |
GenBank mRNA | FQ221080.1 | - | 2 - 3 | 357 | 73 | |
FQ221398.1 | - | 2 - 3 | 357 | 73 | ||
FQ222010.1 | - | 2 - 3 | 360 | 73 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |