Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc5a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013033.2 | NP_037165.1 | 4 - 5 | 543 | 94 | |
GenBank mRNA | AB000729.1 | - | 4 - 5 | 569 | 94 | |
BC081827.1 | - | 4 - 5 | 575 | 7129017 | 94 | |
D16101.1 | - | 4 - 5 | 543 | 94 | ||
U03120.1 | - | 4 - 5 | 501 | 94 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Transporter Mammalian Gene Collection Rattus norvegicus Probe spans exons |