Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eif1ax | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106963.1 | NP_001100433.1 | 4 - 5 | 466 | 133 | |
GenBank mRNA | BC168938.1 | - | 4 - 5 | 339 | 7097069 | 133 |
FQ217925.1 | - | 4 - 5 | 431 | 133 | ||
FQ221854.1 | - | 4 - 5 | 445 | 133 | ||
FQ222873.1 | - | 4 - 5 | 464 | 133 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |