Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Alg3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001142363.1 | NP_001135835.1 | 5 - 6 | 746 | 98 | |
NR_024535.1 | 4 - 5 | 646 | 98 | |||
XM_006248558.2 | XP_006248620.1 | 5 - 6 | 764 | 98 | ||
GenBank mRNA | BC088475.1 | - | 4 - 5 | 629 | 7129249 | 98 |
FM075565.1 | - | 4 - 5 | 558 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus 1700 Inventoried Probe spans exons |