Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spag6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034960.1 | NP_001030132.1 | 3 - 4 | 326 | 104 | |
GenBank mRNA | BC104678.1 | - | 3 - 4 | 326 | 7454069 | 104 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |