Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Etv5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107082.1 | NP_001100552.1 | 5 - 6 | 372 | 77 | |
XM_008768798.1 | XP_008767020.1 | 5 - 6 | 385 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Rattus norvegicus Probe spans exons |