Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017045.1 | NP_058741.1 | 23 - 24 | 2669 | 81 | |
GenBank mRNA | D25233.1 | - | 23 - 24 | 2430 | 81 | |
L07126.1 | - | 2 - 3 | 79 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |