Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dct | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008770941.1 | XP_008769163.1 | 5 - 6 | 1580 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |