Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bin3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013186.1 | NP_001013204.1 | 2 - 3 | 96 | 69 | |
GenBank mRNA | BC079146.1 | - | 2 - 3 | 96 | 7128352 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |