Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bin3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013186.1 | NP_001013204.1 | 4 - 5 | 301 | 80 | |
GenBank mRNA | BC079146.1 | - | 4 - 5 | 301 | 7128352 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |