Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bpifb4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109209.2 | NP_001102679.2 | 4 - 5 | 788 | 60 | |
GenBank mRNA | X60660.1 | - | 2 - 3 | 347 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Probe spans exons |