Search Thermo Fisher Scientific
- Contáctenos
- Orden Rápida
-
¿No tiene una cuenta? Crear una cuenta
Search Thermo Fisher Scientific
Gene Symbol: Actr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031068.1 | NP_112330.1 | 5 - 6 | 512 | 60 | |
GenBank mRNA | AB292042.1 | - | 5 - 6 | 436 | 60 | |
AB292043.1 | - | 5 - 6 | 425 | 60 | ||
BC098014.1 | - | 5 - 6 | 512 | 7454624 | 60 | |
FQ212256.1 | - | 5 - 6 | 714 | 60 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |