Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dapl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108582.1 | NP_001102052.1 | 2 - 3 | 207 | 79 | |
XM_008761859.1 | XP_008760081.1 | 2 - 3 | 396 | 79 | ||
GenBank mRNA | BC168756.1 | - | 2 - 3 | 202 | 6887254 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Apoptosis 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus Probe spans exons |