Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nxf7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037216.1 | NP_001032293.1 | 5 - 6 | 618 | 56 | |
XM_006257254.2 | XP_006257316.1 | 5 - 6 | 651 | 56 | ||
XM_008773431.1 | XP_008771653.1 | 5 - 6 | 1189 | 56 | ||
GenBank mRNA | BC100151.1 | - | 5 - 6 | 618 | 7456178 | 56 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |