Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nxf7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037216.1 | NP_001032293.1 | 7 - 8 | 812 | 104 | |
XM_006257254.2 | XP_006257316.1 | 7 - 8 | 845 | 104 | ||
XM_008773431.1 | XP_008771653.1 | 7 - 8 | 1383 | 104 | ||
GenBank mRNA | BC100151.1 | - | 7 - 8 | 812 | 7456178 | 104 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |