Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wasf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025114.1 | NP_001020285.1 | 2 - 3 | 201 | 131 | |
XM_006256586.2 | XP_006256648.1 | 2 - 3 | 394 | 131 | ||
XM_006256587.2 | XP_006256649.1 | 2 - 3 | 395 | 131 | ||
GenBank mRNA | BC091322.1 | - | 2 - 3 | 201 | 7313558 | 131 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |