Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdh4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008762584.1 | XP_008760806.1 | 8 - 9 | 1425 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |