Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmc7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191950.1 | NP_001178879.1 | 13 - 14 | 1866 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons |