Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wee1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012742.1 | NP_001012760.1 | 1 - 2 | 646 | 87 | |
XM_006229974.2 | XP_006230036.1 | 1 - 2 | 3376 | 87 | ||
GenBank mRNA | BC090346.1 | - | 1 - 2 | 646 | 7306767 | 87 |
D31838.1 | - | 1 - 2 | 845 | 87 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protein Kinase Mammalian Gene Collection Rattus norvegicus Probe spans exons |