Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wee1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012742.1 | NP_001012760.1 | 2 - 3 | 856 | 90 | |
XM_006229974.2 | XP_006230036.1 | 2 - 3 | 3586 | 90 | ||
GenBank mRNA | BC090346.1 | - | 2 - 3 | 856 | 7306767 | 90 |
D31838.1 | - | 2 - 3 | 1055 | 90 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protein Kinase Mammalian Gene Collection Rattus norvegicus Probe spans exons |