Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wee1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012742.1 | NP_001012760.1 | 9 - 10 | 1709 | 68 | |
XM_006229974.2 | XP_006230036.1 | 9 - 10 | 4439 | 68 | ||
GenBank mRNA | BC090346.1 | - | 9 - 10 | 1709 | 7306767 | 68 |
D31838.1 | - | 9 - 10 | 1908 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protein Kinase Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |