Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ovch2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003748959.3 | XP_003749007.1 | 12 - 13 | 1414 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Protease Rattus norvegicus Probe spans exons |