Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nsmce1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039611.1 | NP_001034700.1 | 5 - 6 | 591 | 79 | |
XM_006230291.2 | XP_006230353.1 | 4 - 5 | 433 | 79 | ||
XM_006230292.2 | XP_006230354.1 | 4 - 5 | 1003 | 79 | ||
XM_006230293.2 | XP_006230355.1 | 4 - 5 | 513 | 79 | ||
GenBank mRNA | BC099757.1 | - | 5 - 6 | 591 | 5623851 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |