Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Xylt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006230151.2 | XP_006230213.2 | 8 - 9 | 1675 | 103 | |
GenBank mRNA | AJ295748.1 | - | 5 - 6 | 1181 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |