Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gnb4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013910.2 | NP_001013932.1 | 1 - 2 | 129 | 61 | |
GenBank mRNA | AY552804.1 | - | 1 - 2 | 56 | 61 | |
BC097370.1 | - | 1 - 2 | 129 | 7444022 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Probe spans exons |