Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrc31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006232240.2 | XP_006232302.1 | 2 - 3 | 611 | 76 | |
XM_008760950.1 | XP_008759172.1 | 3 - 4 | 2799 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |