Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Armc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106425.1 | NP_001099895.1 | 5 - 6 | 793 | 61 | |
XM_008760864.1 | XP_008759086.1 | 5 - 6 | 802 | 61 | ||
GenBank mRNA | BC158721.1 | - | 5 - 6 | 784 | 9090461 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |