Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atxn7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006251728.2 | XP_006251790.1 | 11 - 12 | 1813 | 78 | |
XM_008770600.1 | XP_008768822.1 | 11 - 12 | 1813 | 78 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |