Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Antxrl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006252719.2 | XP_006252781.2 | 10 - 11 | 927 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |