Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptpn20b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080149.1 | NP_001073618.1 | 3 - 4 | 316 | 118 | |
XM_006252765.2 | XP_006252827.1 | 4 - 5 | 389 | 118 | ||
XM_006252767.2 | XP_006252829.1 | 4 - 5 | 356 | 118 | ||
XM_008771099.1 | XP_008769321.1 | 4 - 5 | 342 | 118 | ||
XM_008771100.1 | XP_008769322.1 | 4 - 5 | 389 | 118 | ||
GenBank mRNA | BC129117.1 | - | 3 - 4 | 316 | 7114504 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 Protein Phosphatase Mammalian Gene Collection Rattus norvegicus Probe spans exons |