Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grid2ip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105910.1 | NP_001099380.1 | 5 - 6 | 569 | 97 | |
XM_006248865.2 | XP_006248927.1 | 5 - 6 | 639 | 97 | ||
XM_006248866.2 | XP_006248928.1 | 6 - 7 | 1231 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Rattus norvegicus Probe spans exons |