Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Proz | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006253473.1 | XP_006253535.1 | 5 - 6 | 620 | 71 | |
XM_006253474.1 | XP_006253536.1 | 4 - 5 | 506 | 71 | ||
GenBank mRNA | BC160868.1 | - | 5 - 6 | 532 | 71 | |
FQ209855.1 | - | 5 - 6 | 534 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |