Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Upf3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012159.1 | NP_001012159.1 | 4 - 5 | 541 | 61 | |
GenBank mRNA | BC088463.1 | - | 4 - 5 | 541 | 7127783 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |