Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gtpbp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013225.2 | NP_001013243.2 | 4 - 5 | 613 | 60 | |
XM_006244543.2 | XP_006244605.1 | 4 - 5 | 655 | 60 | ||
XM_006244544.2 | XP_006244606.1 | 4 - 5 | 445 | 60 | ||
XM_006244545.2 | XP_006244607.1 | 3 - 4 | 2529 | 60 | ||
XM_006244546.2 | XP_006244608.1 | 4 - 5 | 434 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Probe spans exons |