Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Treml4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006244488.2 | XP_006244550.1 | 1 - 2 | 925 | 118 | |
XM_006244490.2 | XP_006244552.1 | 1 - 2 | 929 | 118 | ||
XM_006244492.2 | XP_006244554.1 | 1 - 2 | 934 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Rattus norvegicus 1700 Probe spans exons |