Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Serpinb9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007732.1 | NP_001007733.1 | 2 - 3 | 335 | 70 | |
XM_006253872.2 | XP_006253934.1 | 2 - 3 | 304 | 70 | ||
GenBank mRNA | BC079063.1 | - | 2 - 3 | 335 | 7121006 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Probe spans exons |